Ascomycin

Supplier: Apollo Scientific
Warning

BIA4317-50MG
APOSBIA4317-50MGEA 59.6 EUR
APOSBIA4317-50MG
Ascomycin
Ascomycin
Formula: C₄₃H₆₉NO₁₂
Storage Temperature: Freezer
MDL Number: MFCD06198665
CAS Number: 104987-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR