4-Nitrophenyl α-D-xylopyranoside

Supplier: Apollo Scientific
Warning

BIB1435-100MG BIB1435-250MG BIB1435-50MG
APOSBIB1435-100MGEA 102 EUR
APOSBIB1435-100MG APOSBIB1435-250MG APOSBIB1435-50MG
4-Nitrophenyl α-D-xylopyranoside
4-Nitrophenyl α-D-xylopyranoside
Formula: C₁₁H₁₃NO₇
Storage Temperature: Freezer
MDL Number: MFCD00067649
CAS Number: 10238-28-5
EINECS: 233-572-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR