D-Glucuronic acid, sodium salt monohydrate

Supplier: Apollo Scientific
Warning

BIB6055-100G BIB6055-250G BIB6055-500G
APOSBIB6055-100GEA 122 EUR
APOSBIB6055-100G APOSBIB6055-250G APOSBIB6055-500G
D-Glucuronic acid, sodium salt monohydrate
D-Glucuronic acid, sodium salt monohydrate
Formula: C₆H₉O₇·Na·H₂O
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00151051
CAS Number: 14984-34-0
EINECS: 239-065-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR