myo-Inositol

Supplier: Apollo Scientific

BIB6083-100G BIB6083-1KG BIB6083-500G
APOSBIB6083-100GEA 36.4 EUR
APOSBIB6083-100G APOSBIB6083-1KG APOSBIB6083-500G
myo-Inositol
myo-Inositol
Formula: C₆H₁₂O₆
Melting Pt: 222…227 °C
Density: 0,63 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
EINECS: 201-781-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR