4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate

Supplier: Apollo Scientific
Warning

BIB6118-1G BIB6118-500MG BIB6118-2.5G
APOSBIB6118-1GEA 125 EUR
APOSBIB6118-1G APOSBIB6118-500MG APOSBIB6118-2.5G
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate
Formula: C₁₀H₇Na₂O₆P·3H₂O
Storage Temperature: Freezer
MDL Number: MFCD00037614
CAS Number: 22919-26-2
EINECS: 245-325-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR