Pyr-βNA

Supplier: Apollo Scientific
Warning

BIB6266-5G BIB6266-1G BIB6266-250MG
APOSBIB6266-5GEA 729 EUR
APOSBIB6266-5G APOSBIB6266-1G APOSBIB6266-250MG
Pyr-βNA
Pyr-βNA
Formula: C₁₅H₁₄N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00055911
CAS Number: 22155-91-5
EINECS: 244-809-9
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR