L(+)-Cysteine

Supplier: Apollo Scientific
Warning

BIC0709-100G BIC0709-1KG BIC0709-250G
APOSBIC0709-100GEA 21.9 EUR
APOSBIC0709-100G APOSBIC0709-1KG APOSBIC0709-250G
L(+)-Cysteine
L-Cysteine
Formula: HSCH₂CH(NH₂)CO₂H
Melting Pt: 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR