Creatine phosphate disodium salt

Supplier: Apollo Scientific
Warning

BIC1452-25G BIC1452-5G
APOSBIC1452-25GEA 522 EUR
APOSBIC1452-25G APOSBIC1452-5G
Creatine phosphate disodium salt
Creatine phosphate disodium salt
Formula: C₄H₈N₃O₅P·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00014351
CAS Number: 6190-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR