SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)

Supplier: Apollo Scientific
Warning

BICL204-100MG
APOSBICL204-100MGEA 190 EUR
APOSBICL204-100MG
SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)
SIAB (N-Succinimidyl (4-iodoacetyl)aminobenzoate)

Primary amine and Sulphhydryl reactiveEnzyme antibody conjugation


Ref: Weltman, J.K.et.al (1983) Bio.Techniques 1, 148-152

Formula: C₁₃H₉IN₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00078537
CAS Number: 72252-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR