2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose ≥99%

Supplier: Apollo Scientific
Warning

BICL4009-10G BICL4009-25G BICL4009-50G
APOSBICL4009-10GEA 422 EUR
APOSBICL4009-10G APOSBICL4009-25G APOSBICL4009-50G
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose ≥99%
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-D-glucopyranose
Formula: C₁₆H₂₃NO₁₀
Melting Pt: 131…133 °C
Storage Temperature: Freezer
MDL Number: MFCD00067367
CAS Number: 14086-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR