N-Butyldeoxynojirimycin 99% min

Supplier: Apollo Scientific

BICL4040-10MG BICL4040-25MG BICL4040-5MG
APOSBICL4040-10MGEA 420 EUR
APOSBICL4040-10MG APOSBICL4040-25MG APOSBICL4040-5MG
N-Butyldeoxynojirimycin 99% min
N-Butyldeoxynojirimycin
Formula: C₁₀H₂₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00272581
CAS Number: 72599-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR