4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside 98% min

Supplier: Apollo Scientific

BICL4164-10G BICL4164-25G BICL4164-5G
APOSBICL4164-10GEA 351 EUR
APOSBICL4164-10G APOSBICL4164-25G APOSBICL4164-5G
4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside 98% min
4-Methoxyphenyl-2,3,4,6-tetra-O-acetyl-beta-D-galactopyranoside
Formula: C₂₁H₂₆O₁₁
Storage Temperature: Ambient
MDL Number: MFCD06797123
CAS Number: 2872-65-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR