5-Acetamidofluorescein di-(β-D-galactopyranoside) ≥95%

Supplier: Apollo Scientific

BICL4232-10MG BICL4232-2MG BICL4232-5MG
APOSBICL4232-10MGEA 1050 EUR
APOSBICL4232-10MG APOSBICL4232-2MG APOSBICL4232-5MG
5-Acetamidofluorescein di-(β-D-galactopyranoside) ≥95%
5-Acetamidofluorescein di-(β-D-galactopyranoside)
Formula: C₃₄H₃₅NO₁₆
Storage Temperature: Ambient
MDL Number: MFCD00467511
CAS Number: 216299-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR