Ruxolitinib phosphate ≥99%

Supplier: Apollo Scientific
Danger

BIFK0028-10MG BIFK0028-25MG BIFK0028-50MG
APOSBIFK0028-10MGEA 65.4 EUR
APOSBIFK0028-10MG APOSBIFK0028-25MG APOSBIFK0028-50MG
Ruxolitinib phosphate ≥99%
Ruxolitinib phosphate

Phosphate Salt, >99%

Formula: C₁₇H₂₁N₆O₄P
Storage Temperature: Freezer
MDL Number: MFCD18452860
CAS Number: 1092939-17-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR