Glycitein

Supplier: Apollo Scientific
Warning

BIG0110-10MG BIG0110-5MG
APOSBIG0110-10MGEA 125 EUR
APOSBIG0110-10MG APOSBIG0110-5MG
Glycitein
Glycitein

Isoflavone derived from Soybean.

Formula: C₁₆H₁₂O₅
Boiling Pt: 547,4 °C
Melting Pt: 330…340 °C
Flash Pt: 210 °C
Storage Temperature: Freezer
MDL Number: MFCD00016679
CAS Number: 40957-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR