L(+)-Glutamic acid ≥99%

Supplier: Apollo Scientific
Warning

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

BIG0707-1KG BIG0707-500G
APOSBIG0707-1KGEA 33.5 EUR
APOSBIG0707-1KG APOSBIG0707-500G
L(+)-Glutamic acid ≥99%
L(+)-Glutamic acid

Ph.Eur, Pyrogen free

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Melting Pt: 205 °C (dec.)
Density: 1,525 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR