D-Glucose-6-phosphate dipotassium salt hydrate

Supplier: Apollo Scientific
Warning

BIG5009-1G BIG5009-5G BIG5009-500MG
APOSBIG5009-1GEA 161 EUR
APOSBIG5009-1G APOSBIG5009-5G APOSBIG5009-500MG
D-Glucose-6-phosphate dipotassium salt hydrate
D-Glucose-6-phosphate dipotassium salt hydrate
Formula: C₆H₁₁K₂O₉P
Storage Temperature: Freezer
CAS Number: 5996-17-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR