Kanamycin sulphate

Supplier: Apollo Scientific
Danger

BIK0126-100G BIK0126-25G BIK0126-250G
APOSBIK0126-100GEA 154 EUR
APOSBIK0126-100G APOSBIK0126-25G APOSBIK0126-250G
Kanamycin sulphate
Kanamycin sulphate

Kanamycin is an aminoglycoside antibiotic and isused as a selective agent for the incorporation of the NPT II (APH3') gene in plant tissue


  • approx. 800 micrograms per mg
  • Cell culture tested

Formula: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00070253
CAS Number: 25389-94-0
EINECS: 246-933-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR