D-(+)-Raffinose pentahydrate

Supplier: Apollo Scientific
Warning

BIK9001-100G BIK9001-1KG BIK9001-500G
APOSBIK9001-100GEA 58 EUR
APOSBIK9001-100G APOSBIK9001-1KG APOSBIK9001-500G
D-(+)-Raffinose pentahydrate
D(+)-Raffinose pentahydrate
Formula: C₁₈H₃₂O₁₆·5H₂O
Storage Temperature: Ambient
MDL Number: MFCD00071590
CAS Number: 17629-30-0
EINECS: 208-146-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR