Melatonin

Supplier: Apollo Scientific
Warning

Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide

BIM0105-1G
APOSBIM0105-1GEA 21.9 EUR
APOSBIM0105-1G
Melatonin
Melatonine
Formula: C₁₃H₁₆N₂O₂
MW: 232.28 g/mol
Melting Pt: 116…118 °C
Storage Temperature: Freezer
MDL Number: MFCD00005655
CAS Number: 73-31-4
EINECS: 200-797-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR