Methotrexate

Supplier: Apollo Scientific
Danger

BIM0130-1G BIM0130-5G BIM0130-10G
APOSBIM0130-1GEA 21.9 EUR
APOSBIM0130-1G APOSBIM0130-5G APOSBIM0130-10G
Methotrexate
Methotrexate

An antineoplastic agent which acts as an antimetaboliteof folic acid.

Formula: C₂₀H₂₂N₈O₅
Melting Pt: 185…204 °C
Storage Temperature: Freezer
MDL Number: MFCD00150847
CAS Number: 59-05-2
UN: 1544
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR