Guanidinium thiocyanate ≥99.0%

Supplier: Apollo Scientific
Danger

BIMB1010-1KG BIMB1010-500G BIMB1010-5KG
APOSBIMB1010-1KGEA 102 EUR
APOSBIMB1010-1KG APOSBIMB1010-500G APOSBIMB1010-5KG
Guanidinium thiocyanate ≥99.0%
Guanidinium thiocyanate

Protein denaturant.


  • Toxic-LD50 of <500mg/Kg

Formula: CH₅N₃·CHNS
Melting Pt: 117…122 °C
Density: 1,29 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR