Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)

Supplier: Apollo Scientific
Warning

Synonyms: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl

BIMI2137-10MG BIMI2137-25MG BIMI2137-5MG
APOSBIMI2137-10MGEA 109 EUR
APOSBIMI2137-10MG APOSBIMI2137-25MG APOSBIMI2137-5MG
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)

Metallo-protease inhibitor selective for aminopeptidase.

Formula: C₁₆H₂₅ClN₂O₄
Melting Pt: 216…218 °C
Storage Temperature: Freezer
MDL Number: MFCD00058004
CAS Number: 65391-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR