2'-Fluoro-2'-deoxyinosine

Supplier: Apollo Scientific
Warning

BIMN2011-100MG BIMN2011-1G BIMN2011-250MG
APOSBIMN2011-100MGEA 102 EUR
APOSBIMN2011-100MG APOSBIMN2011-1G APOSBIMN2011-250MG
2'-Fluoro-2'-deoxyinosine
2'-Fluoro-2'-deoxyinosine
Formula: C₁₀H₁₁FN₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD09750860
CAS Number: 80049-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR