Arabinofuranosyl-isoguanine ≥98%

Supplier: Apollo Scientific

BIMN2041-250MG
APOSBIMN2041-250MGEA 990 EUR
APOSBIMN2041-250MG
Arabinofuranosyl-isoguanine ≥98%
Arabinofuranosyl-isoguanine
Formula: C₁₀H₁₃N₅O₅
Storage Temperature: Ambient
CAS Number: 38819-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR