Nitrofurantoin

Supplier: Apollo Scientific
Danger

Synonyms: 1-[(5-Nitrofurfurylidene)amino]hydantoin

BIN0136-100G BIN0136-250G BIN0136-500G
APOSBIN0136-100GEA 59.6 EUR
APOSBIN0136-100G APOSBIN0136-250G APOSBIN0136-500G
Nitrofurantoin
Nitrofurantoin

Nitrofurantoin is bactericidal in vitro to most gram-positive and gram-negative bacteria.

Formula: C₈H₆N₄O₅
Melting Pt: 267…269 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003224
CAS Number: 67-20-9
EINECS: 200-646-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR