Penicillin V potassium salt

Supplier: Apollo Scientific
Danger

Synonyms: Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin V Potassium

BIP0143-25G BIP0143-5G
APOSBIP0143-25GEA 116 EUR
APOSBIP0143-25G APOSBIP0143-5G
Penicillin V potassium salt
Penicillin V potassium salt

Penicillin V is an inhibitor of bacterial cell wall synthesisActive against gram-positive and some gram-negative bacteria. b-lactamase sensitive.


  • Potency: 1650mg/mg

Formula: C₁₆H₁₇KN₂O₅S
Melting Pt: 197…202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051771
CAS Number: 132-98-9
EINECS: 205-086-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR