Azido-PEG24-alcohol

Supplier: Apollo Scientific
Warning

BIPG1120-100MG BIPG1120-250MG
APOSBIPG1120-100MGEA 364 EUR
APOSBIPG1120-100MG APOSBIPG1120-250MG
Azido-PEG24-alcohol
Azido-PEG24-alcohol
Formula: C₄₈H₉₇N₃O₂₄
Storage Temperature: Refrigerator
CAS Number: 73342-16-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR