Fmoc-N-amido-PEG10-Acid

Supplier: Apollo Scientific

BIPG1404-100MG
APOSBIPG1404-100MGEA 21.9 EUR
APOSBIPG1404-100MG
Fmoc-N-amido-PEG10-Acid
Fmoc-N-amido-PEG10-Acid
Formula: C₃₈H₅₇NO₁₄
Storage Temperature: Ambient
MDL Number: MFCD28122954
CAS Number: 2101563-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR