D-(+)-Sucrose

Supplier: Apollo Scientific
Warning

Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

BIS3391-1KG BIS3391-500G
APOSBIS3391-1KGEA 27.7 EUR
APOSBIS3391-1KG APOSBIS3391-500G
D-(+)-Sucrose
D(+)-Sucrose

  • Plant tissue culture tested

Formula: C₁₂H₂₂O₁₁
Melting Pt: 185…190 °C
Density: 1,58 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR