Sinapic acid 98%

Supplier: Apollo Scientific
Warning

BIS8112-5G BIS8112-25G BIS8112-100G
APOSBIS8112-5GEA 21.9 EUR
APOSBIS8112-5G APOSBIS8112-25G APOSBIS8112-100G
Sinapic acid 98%
Sinapic acid

  • Protein

Formula: C₁₁H₁₂O₅
Melting Pt: 195…202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004401
CAS Number: 530-59-6
EINECS: 208-487-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR