TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98%

Supplier: Apollo Scientific
Danger

Synonyms: 3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride

BIT0122-10G BIT0122-25G BIT0122-5G
APOSBIT0122-10GEA 125 EUR
APOSBIT0122-10G APOSBIT0122-25G APOSBIT0122-5G
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98%
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

Used in the reduction of disulphide groups.


  • Water soluble
  • More stablethan DTT

Formula: C₉H₁₆ClO₆P
MW: 286.65 g/mol
Melting Pt: 177…178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145469
CAS Number: 51805-45-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR