TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt) ≥98%

Supplier: Apollo Scientific
Warning

BIT3002-100G
APOSBIT3002-100GEA 104 EUR
APOSBIT3002-100G
TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt) ≥98%
TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt)

  • Buffer

Formula: C₇H₁₆NNaO₆S
Storage Temperature: Ambient
MDL Number: MFCD00069903
CAS Number: 91000-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR