Human Recombinant IL-2

Supplier: Apollo Scientific

BITP1126-1MG
APOSBITP1126-1MGEA 932 EUR
APOSBITP1126-1MG
Human Recombinant IL-2
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name IL-2
Environmentally Preferable
Protein synonyms Interleukin-2
Protein/peptide type Recombinant
Species Human
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR