Sheep Recombinant Leptin tA

Supplier: Apollo Scientific

BITP1209-1MG
APOSBITP1209-1MGEA 0 EUR
APOSBITP1209-1MG
Sheep Recombinant Leptin tA
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name Leptin tA
Environmentally Preferable
Protein synonyms Leptin Triple Antagonist
Protein/peptide type Recombinant
Species Sheep
Source

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR