Anti-Hepatitis B Surface Antigen A Goat Polyclonal Antibody

Supplier: Apollo Scientific
BITP2043-10ML BITP2043-1ML BITP2043-5ML
APOSBITP2043-10MLEA 0 EUR
APOSBITP2043-10ML APOSBITP2043-1ML APOSBITP2043-5ML
Anti-Hepatitis B Surface Antigen A Goat Polyclonal Antibody
Antibodies
Order Now

SPECIFICATIONS

Antigen name Hepatitis B Surface Antigen A
Environmentally Preferable
Clonality Polyclonal
Reactivity
Host Goat
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR