Bismuth(III) oxide 99,9%

Supplier: Apollo Scientific
Warning

IN1252-100G IN1252-500G
APOSIN1252-100GEA 190 EUR
APOSIN1252-100G APOSIN1252-500G
Bismuth(III) oxide 99,9%
Bismuth (III) oxide
Formula: BiO₃
Boiling Pt: 1890 °C
Melting Pt: 817 °C
Density: 8,9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR