Copper(II) oxide 99,9%

Supplier: Apollo Scientific
Warning

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

IN1576-500G
APOSIN1576-500GEA 415 EUR
APOSIN1576-500G
Copper(II) oxide 99,9%
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1362 °C
Density: 6.315 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
UN: 3077
ADR: 9+EHS III
Merck Index: 13,02674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR