Dysprosium(III) oxide 99,9%

Supplier: Apollo Scientific
Warning

Synonyms: Didysprosium trioxide, Dysprosium oxide

IN1663-100G IN1663-250G IN1663-25G
APOSIN1663-100GEA 618 EUR
APOSIN1663-100G APOSIN1663-250G APOSIN1663-25G
Dysprosium(III) oxide 99,9%
Dysprosium (III) oxide
Formula: Dy₂O₃
Melting Pt: 2340 °C
Density: 7,81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010986
CAS Number: 1308-87-8
EINECS: 215-164-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR