Holmium(III) oxide 99,9%

Supplier: Apollo Scientific
Warning

Synonyms: Diholmium trioxide, Holmium oxide

IN1990-50G
APOSIN1990-50GEA 277 EUR
APOSIN1990-50G
Holmium(III) oxide 99,9%
Holmium (III) oxide
Formula: Ho₂O₃
Boiling Pt: 3900 °C
Melting Pt: 2360 °C
Density: 8,36 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011053
CAS Number: 12055-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR