Neodymium trichloride hexahydrate 99,9%

Supplier: Apollo Scientific
Warning

Synonyms: Neodymium (III) chloride hexahydrate

IN2632-100G IN2632-50G
APOSIN2632-100GEA 248 EUR
APOSIN2632-100G APOSIN2632-50G
Neodymium trichloride hexahydrate 99,9%
Neodymium trichloride hexahydrate
Formula: Cl₃Nd·6H₂O
Melting Pt: 124 °C
Density: 2,282 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149801
CAS Number: 13477-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR