Oxalic acid dihydrate ACS

Supplier: Apollo Scientific
Danger

Synonyms: Ethanedioic acid dihydrate

IN2794-1KG
APOSIN2794-1KGEA 190 EUR
APOSIN2794-1KG
Oxalic acid dihydrate ACS
Oxalic acid dihydrate
Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Melting Pt: 101…102 °C
Density: 1.65 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 607-634-3
UN: 3261
ADR: 8 III
Merck Index: 13,06980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR