Diphosphorus pentoxide 98%

Supplier: Apollo Scientific
Danger

Synonyms: Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride

IN2857-2.5KG IN2857-500G
APOSIN2857-2.5KGEA 177 EUR
APOSIN2857-2.5KG APOSIN2857-500G
Diphosphorus pentoxide 98%
di-Phosphorus pentoxide
Formula: P₂O₅
Melting Pt: 340 °C (subl.)
Density: 2,39 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR