Thulium(III) oxide 99.99%

Supplier: Apollo Scientific
Warning

IN3616-1G IN3616-5G
APOSIN3616-1GEA 125 EUR
APOSIN3616-1G APOSIN3616-5G
Thulium(III) oxide 99.99%
Thulium (III) oxide
Formula: O₃Tm₂
Melting Pt: 2425 °C
Storage Temperature: Ambient
MDL Number: MFCD00011285
CAS Number: 12036-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR