Triphenylphosphine polymer bound 1,4 - 2,0 mmol/g on polystyrene, cross-linked with 1% DVB 200 - 400 mesh

Supplier: Apollo Scientific
Warning

Synonyms: Triphenylphosphine resin

IN3722-5G IN3722-25G IN3722-100G
APOSIN3722-5GEA 50.9 EUR
APOSIN3722-5G APOSIN3722-25G APOSIN3722-100G
Triphenylphosphine polymer bound 1,4 - 2,0 mmol/g on polystyrene, cross-linked with 1% DVB 200 - 400 mesh
Triphenylphosphine polymer bound
Formula: C₁₈H₁₅P
MW: 262.292 g/mol
MDL Number: MFCD20489348
CAS Number: 39319-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR