a-Tosyl(4-bromobenzyl)isocyanide 97%

Supplier: Apollo Scientific
Warning

OR01701-1G OR01701-250MG
APOSOR01701-1GEA 325 EUR
APOSOR01701-1G APOSOR01701-250MG
a-Tosyl(4-bromobenzyl)isocyanide 97%
a-Tosyl(4-bromobenzyl)isocyanide
Formula: C₁₅H₁₂BrNO₂S
Storage Temperature: Ambient
CAS Number: 655254-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR