1-Methyl-1H-indazol-3-amine

Supplier: Apollo Scientific

OR01970-1G OR01970-250MG OR01970-5G
APOSOR01970-1GEA 27.7 EUR
APOSOR01970-1G APOSOR01970-250MG APOSOR01970-5G
1-Methyl-1H-indazol-3-amine
1-Methyl-1H-indazol-3-amine
Formula: C₈H₉N₃
MDL Number: MFCD09054755
CAS Number: 60301-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR