(Cyclohexylmethyl)hydrazine hydrochloride 97%

Supplier: Apollo Scientific
Warning

Synonyms: (Hydrazinomethyl)cyclohexane hydrochloride

OR0199-1G OR0199-5G OR0199-10G
APOSOR0199-1GEA 102 EUR
APOSOR0199-1G APOSOR0199-5G APOSOR0199-10G
(Cyclohexylmethyl)hydrazine hydrochloride 97%
(Cyclohexylmethyl)hydrazine hydrochloride
Formula: C₇H₁₆N₂·ClH
Melting Pt: 141…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06245509
CAS Number: 3637-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR