1-(5-Chloropyridin-2-yl)-1,4-diazepane

Supplier: Apollo Scientific
Warning

OR01997-1G OR01997-5G
APOSOR01997-1GEA 198 EUR
APOSOR01997-1G APOSOR01997-5G
1-(5-Chloropyridin-2-yl)-1,4-diazepane
1-(5-Chloropyridin-2-yl)-1,4-diazepane
Formula: C₁₀H₁₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD05182308
CAS Number: 502133-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR