4-Chlorophenyl methyl sulphone 98%

Supplier: Apollo Scientific
Warning

OR0678-100G OR0678-10G OR0678-50G
APOSOR0678-100GEA 98.6 EUR
APOSOR0678-100G APOSOR0678-10G APOSOR0678-50G
4-Chlorophenyl methyl sulphone 98%
4-Chlorophenyl methyl sulfone
Formula: C₇H₇ClO₂S
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00025067
CAS Number: 98-57-7
EINECS: 202-682-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR